751863
Scandium(III) oxide
99.99% trace rare earth metals basis
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Recommended Products
Assay
99.99% trace rare earth metals basis
form
powder
application(s)
battery manufacturing
SMILES string
O=[Sc]O[Sc]=O
InChI
1S/3O.2Sc
InChI key
HYXGAEYDKFCVMU-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Suitable for vacuum deposition applications
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Laser Physics Letters, 5, 109-109 (2008)
Physica Status Solidi, 188, 239-239 (2001)
The journal of physical chemistry. B, 109(39), 18324-18331 (2006-07-21)
Nanostructured scandium hydrous oxides were hydrothermally synthesized at 180 degrees C for 18 h, using NaOH, NH(4)OH, and KOH as the bases. They were characterized by means of X-ray diffraction (XRD), transmission electron microscopy (TEM), N2 adsorption, thermogravimetry and differential
The journal of physical chemistry. A, 111(45), 11572-11578 (2007-10-18)
The reactions of scandium atoms and O(2) have been reinvestigated using matrix isolation infrared spectroscopy and density functional theory calculations. A series of new oxygen-rich scandium oxide/dioxygen complexes were prepared and characterized. The ground state scandium atoms react with dioxygen
Optics express, 18(11), 11173-11180 (2010-07-01)
We report on the measurements of near-UV excited-state absorption (ESA) spectra and refractive index changes (RICs) in the two ytterbium doped laser crystals Yb:Lu2O3 and Yb:Sc2O3. ESA is assigned to ligand-to-metal charge transfer (LMCT) absorption transitions and RICs to the
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service