Skip to Content
Merck
All Photos(1)

Key Documents

751332

Sigma-Aldrich

3-epi-25-Hydroxyvitamin D3 (6,19,19-d3) solution

100 μg/mL in ethanol, 98 atom % D, 98% (CP)

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
$1,630.00

$1,630.00


Check Cart for Availability

Request more information

Select a Size

Change View
1 ML
$1,630.00

About This Item

Empirical Formula (Hill Notation):
C27D3H41O2
Molecular Weight:
403.66
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.24

$1,630.00


Check Cart for Availability

Request more information

isotopic purity

98 atom % D

Quality Level

Assay

98% (CP)

form

solution

concentration

100 μg/mL in ethanol

technique(s)

mass spectrometry (MS): suitable

color

colorless

application(s)

detection

mass shift

M+3

storage temp.

−20°C

SMILES string

[2H]/C([2H])=C1CC[C@@H](O)CC\1=C([2H])\C=C2/CCC[C@]3(C)[C@H](CC[C@@H]23)[C@H](C)CCCC(C)(C)O

InChI

1S/C27H44O2/c1-19-10-13-23(28)18-22(19)12-11-21-9-7-17-27(5)24(14-15-25(21)27)20(2)8-6-16-26(3,4)29/h11-12,20,23-25,28-29H,1,6-10,13-18H2,2-5H3/b21-11+,22-12-/t20-,23-,24-,25+,27-/m1/s1/i1D2,12D

InChI key

JWUBBDSIWDLEOM-BOFGWHLFSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

3-epi-25-Hydroxyvitamin D3 [3-epi-25(OH)D3] or (C3-epimer) is a metabolite of vitamin D. It is an epimer of 25-hydroxyvitamin D3 which differs only at C-3 position wherein -OH group at C-3 carbon is in the α orientation rather than in the β orientation.[1]
3-epi-25-Hydroxyvitamin D3 (6,19,19-d3) is a deuterated vitamin D3 wherein C-6 and C-19 protons are replaced by deuterium.

Application

3-epi-25-Hydroxyvitamin D3 (6,19,19-d3) can be used as an analytical standard for the quantitative analysis of 3-epi-25-hydroxyvitamin D3 content in biological fluids via LC-MS/MS.[1]

Packaging

1 mL in ampule

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

57.2 °F - closed cup - Solvent

Flash Point(C)

14.0 °C - closed cup - Solvent


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Overestimation of 25-hydroxyvitamin D3 by increased ionisation efficiency of 3-epi-25-hydroxyvitamin D3 in LC-MS/MS methods not separating both metabolites as determined by an LC-MS/MS method for separate quantification of 25-hydroxyvitamin D3, 3-epi-25-hydroxyvitamin D3 and 25-hydroxyvitamin D2 in human serum
van den Ouweland JMW, et al.
Journal of Chromatography. B, Analytical Technologies in the Biomedical and Life Sciences, 967, 195-202 (2014)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service