Skip to Content
Merck
All Photos(1)

Key Documents

750255

Sigma-Aldrich

Potassium 4-dibenzothiophenetrifluoroborate

97%

Synonym(s):

Potassium 4-dibenzothienyltrifluoroborate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H7BF3KS
CAS Number:
Molecular Weight:
290.15
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

289-294 °C

SMILES string

[K+].F[B-](F)(F)c1cccc2c3ccccc3sc12

InChI

1S/C12H7BF3S.K/c14-13(15,16)10-6-3-5-9-8-4-1-2-7-11(8)17-12(9)10;/h1-7H;/q-1;+1

InChI key

XYTNZQSGZIYGCR-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service