74814
1,1,3,3,5,5,7,7-Octamethyltetrasiloxane
technical, ≥85% (GC)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C8H26O3Si4
CAS Number:
Molecular Weight:
282.63
Beilstein:
1770063
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
Recommended Products
grade
technical
Assay
≥85% (GC)
refractive index
n20/D 1.388 (lit.)
bp
169 °C (lit.)
density
0.862 g/mL at 20 °C (lit.)
SMILES string
C[SiH](C)O[Si](C)(C)O[Si](C)(C)O[SiH](C)C
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service