Skip to Content
Merck
All Photos(3)

Key Documents

747092

Sigma-Aldrich

C8-BTBT

≥99% (HPLC)

Synonym(s):

2,7-Dioctyl[1]benzothieno[3,2-b][1]benzothiophene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H40S2
CAS Number:
Molecular Weight:
464.77
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

≥99% (HPLC)

form

powder

mp

108-112 °C

semiconductor properties

P-type (mobility=5.5 cm2/V·s)

SMILES string

CCCCCCCCC1=CC2=C(C=C1)C3=C(C(C=CC(CCCCCCCC)=C4)=C4S3)S2

InChI

1S/C30H40S2/c1-3-5-7-9-11-13-15-23-17-19-25-27(21-23)31-30-26-20-18-24(22-28(26)32-29(25)30)16-14-12-10-8-6-4-2/h17-22H,3-16H2,1-2H3

InChI key

YWIGIVGUASXDPK-UHFFFAOYSA-N

General description

C8-BTBT is a conducting polymer with [1]benzothieno[3,2-b][1]- benzothiophene (BTBT) as the base material for the development of air-stable semiconductors. It can form a spin coated thin film that can be used as a p-type semiconductor with charge mobility of 43 cm2V-1s-1. It exhibits highly ordered self-assembled monolayer (SAM) on graphene and hexagonal boron nitride (hBN) [1][2][3] with sheet resistance: <1,000 ohm/sq

Application

C8-BTBT is a conjugating polymer that can be used in a variety of organic electronics based applications which includes organic photovoltaic cells (OPV), organic light emitting diodes (OLEDs) and organic thin film transistors (OTFTs).[4][5][6]
High field-effect mobility of 5.5-5.7 cm2/Vs and high On/Off ratio of 109 [7]; solution-processed (solubility of 80 mg/mL) OFETs

Legal Information

Product of Nippon Kayaku

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jeong-Wan Jo et al.
Materials (Basel, Switzerland), 13(23) (2020-12-11)
The development of novel dielectric materials with reliable dielectric properties and low-temperature processibility is crucial to manufacturing flexible and high-performance organic thin-film transistors (OTFTs) for next-generation roll-to-roll organic electronics. Here, we investigate the solution-based fabrication of high-k aluminum oxide (Al2O3)
Lu Lyu et al.
Physical chemistry chemical physics : PCCP, 19(2), 1669-1676 (2016-12-21)
Combining ultraviolet photoemission spectroscopy (UPS), X-ray photoemission spectroscopy (XPS), atomic force microscopy (AFM) and small angle X-ray diffraction (SAXD) measurements, we perform a systematic investigation on the correlations of the electronic structure, film growth and molecular orientation of 2,7-diocty[1]benzothieno[3,2-b]benzothiophene (C8-BTBT)
Ultrahigh Vacuum Self-Assembly of Rotationally Commensurate C8-BTBT/MoS2/Graphene Mixed-Dimensional Heterostructures.
Liu X, et al.
Chemistry of Materials, 23(4), 523-526 (2019)
Zhimin Chai et al.
Nanotechnology, 30(48), 485203-485203 (2019-08-31)
Achieving low-cost fabrication of organic field-effect transistors (OFETs) has long been pursued in the semiconductor industry. Solution-based process allows the fabrication of OFETs cost-effective because of its merit of vacuum-free and room temperature operation. Here, we show a facile and
Darmawan; Peter;
Advances in Functional Materials, 22(21), 4577-4583 (2012)

Articles

Small molecular weight organic semiconductors are promising for flexible transistor applications in next-gen soft electronics.

Intrinsically stretchable active layers for organic field-effect transistors (OFET) are discussed. Polymer structural modification & post-polymerization modifications are 2 methods to achieve this.

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service