Skip to Content
Merck
All Photos(2)

Key Documents

746835

Sigma-Aldrich

Iron

nanopowder, 25 nm avg. part. size, 99.5% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Fe
CAS Number:
Molecular Weight:
55.85
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Assay

99.5% trace metals basis

form

nanopowder

resistivity

9.71 μΩ-cm

avg. part. size

25 nm

bp

2750 °C (lit.)

mp

1535 °C (lit.)

density

7.86 g/mL at 25 °C (lit.)

application(s)

battery manufacturing

SMILES string

[Fe]

InChI

1S/Fe

InChI key

XEEYBQQBJWHFJM-UHFFFAOYSA-N

Application

Iron nanoparticles have been investigated for analytical applications including water purification of arsenic, organic contaminants, and magnetic nanocomposites, as well as biomedical applications.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Sol. 2 - Self-heat. 2

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

nwg

Flash Point(F)

69.8 °F

Flash Point(C)

21 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Song; Hocheol;
Applied Catalysis. B, Environmental, 78(1-2), 53-60 (2008)
Sushil Raj Kanel et al.
Environmental science & technology, 39(5), 1291-1298 (2005-03-25)
Nanoscale zero-valent iron (NZVI) was synthesized and tested for the removal of As(III), which is a highly toxic, mobile, and predominant arsenic species in anoxic groundwater. We used SEM-EDX, AFM, and XRD to characterize particle size, surface morphology, and corrosion
Wilson;J. L.;
Journal of Applied Physics, 95(3), 1439-1443 (2004)
Kaori Kohara et al.
Chemical communications (Cambridge, England), 49(25), 2563-2565 (2013-02-21)
Carboxylated SiO2-coated α-Fe nanoparticles have been successfully prepared via CaH2-mediated reduction of SiO2-coated Fe3O4 nanoparticles followed by surface carboxylation. These α-Fe-based nanoparticles, which are characterized by ease of coating with additional functional groups, a large magnetization of 154 emu per

Articles

Professor Hui Mao explores the use of superparamagnetic iron oxide nanoparticles (INOPs) that offer an alternate contrast-enhancing mechanism.

TiO2 exhibits wide band gap semiconductor and memristor properties electronically, with high opacity and UV absorbance optically.

Prof. Yadong Yin discusses various synthesis methods of magnetite nanocrystals and their applications in different fields.

Magnetic materials find diverse applications from data storage to renewable energy.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service