Skip to Content
Merck
All Photos(2)

Key Documents

746118

Sigma-Aldrich

Sodium difluoroheptylazidosulfinate

95%

Synonym(s):

Baran DAAS-Na Reagent, Sodium 1,1-difluoro-7-azido-heptylsulfinate, Sodium 7-azido-1,1-difluoroheptanesulfinate

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$342.00

$342.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
$342.00

About This Item

Empirical Formula (Hill Notation):
C7H12F2N3NaO2S
CAS Number:
Molecular Weight:
263.24
MDL number:
UNSPSC Code:
12352125
PubChem Substance ID:
NACRES:
NA.22

$342.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

95%

form

solid

reaction suitability

reaction type: C-C Bond Formation
reagent type: catalyst
reaction type: C-H Activation

reagent type: linker

functional group

azide
fluoro
sulfinic acid

storage temp.

2-8°C

SMILES string

[Na+].[O-]S(=O)C(F)(F)CCCCCCN=[N+]=[N-]

InChI

1S/C7H13F2N3O2S.Na/c8-7(9,15(13)14)5-3-1-2-4-6-11-12-10;/h1-6H2,(H,13,14);/q;+1/p-1

InChI key

FFNHIEJQAMGBCZ-UHFFFAOYSA-M

Application

Within heteroarene platforms, DAAS-Na allows for the direct introduction of a difluoroalkyl chain carrying an azide group, which is designed for the purpose of bioconjugation. Through a Huisgen cycloaddition (a "click" reaction), the azide moiety can then be reacted with a terminal alkyne that is bound to a functional molecule such as biomacromolecules. This effectively allows for a covalent linkage between a heteroarene and any target molecule, for example, between a heteroarene-containing drug and an antibody. This reagent was developed by the Baran Group and is one of several reagents for the direct alkylation of heterocycles.

Learn More at the Professor and Product Portal of Professor Phil S. Baran.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qianghui Zhou et al.
Journal of the American Chemical Society, 135(35), 12994-12997 (2013-08-21)
A general C-H functionalization method for the tagging of natural products and pharmaceuticals is described. An azide-containing sulfinate reagent allows the appendage of azidoalkyl chains onto heteroaromatics, the product of which can then be attached to a monoclonal antibody by
Direct synthesis of fluorinated heteroarylether bioisosteres.
Qianghui Zhou et al.
Angewandte Chemie (International ed. in English), 52(14), 3949-3952 (2013-03-06)
Fionn O'Hara et al.
Journal of the American Chemical Society, 135(32), 12122-12134 (2013-07-19)
Radical addition processes can be ideally suited for the direct functionalization of heteroaromatic bases, yet these processes are only sparsely used due to the perception of poor or unreliable control of regiochemistry. A systematic investigation of factors affecting the regiochemistry

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service