Skip to Content
Merck
All Photos(1)

Documents

744956

Sigma-Aldrich

2-(Tosyloxy)cycloheptanone

≥98% (HPLC)

Synonym(s):

α-Tosyloxycycloheptanone, 2-Hydroxycycloheptanone p-toluenesulfonate, 2-{[(4-Methylphenyl)sulfonyl]oxy}cycloheptanone, Toluene-4-sulfonic acid 2-oxocycloheptyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H18O4S
CAS Number:
Molecular Weight:
282.36
Beilstein:
5820980
UNSPSC Code:
12352108
PubChem Substance ID:

Quality Level

Assay

≥98% (HPLC)

form

solid

reaction suitability

reaction type: C-C Bond Formation

SMILES string

O=C1CCCCCC1OS(=O)(C2=CC=C(C)C=C2)=O

InChI

1S/C14H18O4S/c1-11-7-9-12(10-8-11)19(16,17)18-14-6-4-2-3-5-13(14)15/h7-10,14H,2-6H2,1H3

InChI key

WVZOAZUIYHADSM-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

m-Iodosylbenzoic Acid: Recyclable Hypervalent Iodine Reagent for ?-Tosyloxylation and ?-Mesyloxylation of Ketones
Yusubov, M.S., et al.
Synthetic Communications, 39, 3772-3784 (2009)
Reaction of [hydroxy(tosyloxy)iodo]benzene and [hydroxy(mesyloxy)iodo]benzene with trimethylsilyl enol ethers. A new general method for .alpha.-sulfonyloxylation of carbonyl compounds.
Moriarty, R. M., et al.
The Journal of Organic Chemistry, 54, 1101-1104 (1989)
One-step .alpha.-tosyloxylation of ketones with [hydroxy(tosyloxy)iodo]benzene
Koser, G.F., et al.
The Journal of Organic Chemistry, 47, 2487-2489 (1982)
Scott T Handy et al.
The Journal of organic chemistry, 70(7), 2874-2877 (2005-03-25)
[structure: see text] We report a homogeneous supported version of Koser's salt based on a room-temperature ionic liquid (RTIL) support. By altering the nature of the RTIL, a material was developed that was stable, recyclable, and readily separable from the
Makoto Ueno et al.
The Journal of organic chemistry, 68(16), 6424-6426 (2003-08-05)
alpha-Tosyloxyketones and alpha-tosyloxyaldehydes were directly prepared from alcohols by treatment with iodosylbenzene and p-toluenesulfonic acid monohydrate in good yields. This method can be used for the direct preparation of thiazoles, imidazoles, and imidazo[1,2-a]pyridines from alcohols in good to moderate yields

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service