Skip to Content
Merck
All Photos(1)

Key Documents

744212

Sigma-Aldrich

Diethyl carbamoylmethylphosphonate

95%

Synonym(s):

Diethyl 2-amino-2-oxoethylphosphonate, Diethyl aminocarbonylmethylphosphonate

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$69.30

$69.30


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$69.30

About This Item

Empirical Formula (Hill Notation):
C6H14NO4P
CAS Number:
Molecular Weight:
195.15
Beilstein:
1777584
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

$69.30


Check Cart for Availability

Request a Bulk Order

Assay

≥95% (CHN)
95%

form

powder

composition

carbon, 34.9-38.9%
nitrogen, 6.8-7.6%

reaction suitability

reaction type: C-C Bond Formation

functional group

amide
phosphonate

SMILES string

CCOP(=O)(CC(N)=O)OCC

InChI

1S/C6H14NO4P/c1-3-10-12(9,11-4-2)5-6(7)8/h3-5H2,1-2H3,(H2,7,8)

InChI key

QPVCUQOSWAXEOQ-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service