Skip to Content
Merck
All Photos(1)

Documents

743747

Sigma-Aldrich

(3-Bromophenyl)(2,4,6-trimethylphenyl)iodonium triflate

≥95%

Synonym(s):

(3-Bromophenyl)(2,4,6-trimethylphenyl)iodonium trifluoromethanesulfonate, (3-Bromophenyl)mesityliodonium triflate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H15BrF3IO3S
CAS Number:
Molecular Weight:
551.16
Beilstein:
20041340
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

Assay

≥95% (HPLC)
≥95%

form

powder

composition

carbon content, 33.0-36.8%

SMILES string

[O-]S(=O)(=O)C(F)(F)F.Cc1cc(C)c([I+]c2cccc(Br)c2)c(C)c1

InChI

1S/C15H15BrI.CHF3O3S/c1-10-7-11(2)15(12(3)8-10)17-14-6-4-5-13(16)9-14;2-1(3,4)8(5,6)7/h4-9H,1-3H3;(H,5,6,7)/q+1;/p-1

InChI key

QTQFCDQRJRJUAN-UHFFFAOYSA-M

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service