Skip to Content
Merck
All Photos(1)

Documents

743097

Sigma-Aldrich

Bis(4-fluorophenyl)iodonium triflate

≥98% (HPLC)

Synonym(s):

(p-F-Ph)2IOTf, Bis(4-fluorophenyl)iodonium trifluoromethanesulfonate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H8F5IO3S
CAS Number:
Molecular Weight:
466.16
Beilstein:
10620586
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:

Assay

≥98% (HPLC)

form

powder

composition

carbon, 32.7-34.3% CHN

reaction suitability

reagent type: oxidant

SMILES string

[O-]S(=O)(=O)C(F)(F)F.Fc1ccc([I+]c2ccc(F)cc2)cc1

InChI

1S/C12H8F2I.CHF3O3S/c13-9-1-5-11(6-2-9)15-12-7-3-10(14)4-8-12;2-1(3,4)8(5,6)7/h1-8H;(H,5,6,7)/q+1;/p-1

InChI key

HTFNIUDXMGKTQK-UHFFFAOYSA-M

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maart van Druenen et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 26(72), 17581-17587 (2020-10-03)
Black phosphorus (BP) has extraordinary properties, but its ambient instability remains a critical challenge. Functionalization has been employed to overcome the sensitivity of BP to ambient conditions while preserving its properties. Herein, a simultaneous exfoliation-functionalization process is reported that functionalizes

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service