Skip to Content
Merck
All Photos(1)

Key Documents

742279

Sigma-Aldrich

(2S,2′S)-2,2′-Bipyrrolidine

≥99.0% (GC)

Synonym(s):

(S,S)-2,2′-Bipyrrolidine, (S,S)-2,2′-Bipyrrolidinyl

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
$146.00
2 G
$503.00

$146.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 MG
$146.00
2 G
$503.00

About This Item

Empirical Formula (Hill Notation):
C8H16N2
CAS Number:
Molecular Weight:
140.23
Beilstein:
5376256
MDL number:
UNSPSC Code:
12161600
NACRES:
NA.22

$146.00


Check Cart for Availability

Request a Bulk Order

Assay

≥99.0% (GC)

form

powder

optical purity

enantiomeric excess: ≥99.5% (GC)

reaction suitability

reagent type: ligand

storage temp.

2-8°C

SMILES string

N1C(CCC1)C2NCCC2

InChI

1S/C8H16N2/c1-3-7(9-5-1)8-4-2-6-10-8/h7-10H,1-6H2

InChI key

NQHVTVSAFRAXPA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service