Skip to Content
Merck
All Photos(1)

Key Documents

740241

Sigma-Aldrich

1-(Boc-amino)-3-butene

97%

Synonym(s):

tert-Butyl but-3-enylcarbamate, Boc-4-amino-1-butene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H17NO2
CAS Number:
Molecular Weight:
171.24
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
form:
liquid
Assay:
97%

Quality Level

Assay

97%

form

liquid

refractive index

n20/D 1.443

density

0.929 g/mL at 25 °C

functional group

allyl
amine

storage temp.

2-8°C

SMILES string

CC(C)(C)OC(=O)NCCC=C

InChI

1S/C9H17NO2/c1-5-6-7-10-8(11)12-9(2,3)4/h5H,1,6-7H2,2-4H3,(H,10,11)

InChI key

YWSXTMBDIBZHBB-UHFFFAOYSA-N

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

213.8 °F

Flash Point(C)

101 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Betar M Gallant et al.
ACS nano, 9(5), 5143-5153 (2015-04-16)
The interfacial shear strength between Si microwires and a Nafion membrane has been tailored through surface functionalization of the Si. Acidic (-COOH-terminated) or basic (-NH2-terminated) surface-bound functionality was introduced by hydrosilylation reactions to probe the interactions between the functionalized Si

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service