Skip to Content
Merck
All Photos(1)

Documents

739006

Sigma-Aldrich

(2-Bromophenyl)dicyclohexylphosphine

97%

Synonym(s):

2-Bromo-dicyclohexylphenylphosphine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H26BrP
CAS Number:
Molecular Weight:
353.28
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

solid

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand

mp

76-79 °C

functional group

phosphine

SMILES string

Brc1ccccc1P(C2CCCCC2)C3CCCCC3

InChI

1S/C18H26BrP/c19-17-13-7-8-14-18(17)20(15-9-3-1-4-10-15)16-11-5-2-6-12-16/h7-8,13-16H,1-6,9-12H2

InChI key

QFPIIICNJHDAKC-UHFFFAOYSA-N

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service