Skip to Content
Merck
All Photos(2)

Key Documents

738999

Sigma-Aldrich

Isooctyl trimethoxysilane, mixture of isomers

97%

Synonym(s):

2,4,4-Trimethylpentyltrimethoxysilane

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$98.00
100 G
$333.00

$98.00


Available to ship on14 April 2025Details


Request a Bulk Order

Select a Size

Change View
25 G
$98.00
100 G
$333.00

About This Item

Linear Formula:
C8H17Si(OCH3)3
CAS Number:
Molecular Weight:
234.41
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.22

$98.00


Available to ship on14 April 2025Details


Request a Bulk Order

Quality Level

Assay

97%

form

liquid

refractive index

n20/D 1.416

density

0.901 g/mL at 25 °C

storage temp.

2-8°C

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

125.6 °F

Flash Point(C)

52 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service