Skip to Content
Merck
All Photos(1)

Key Documents

738638

Sigma-Aldrich

Dichloro(hexamethylbenzene)ruthenium(II) dimer

Synonym(s):

(Hexamethylbenzene)Ru(II)Cl2 dimer, [(C6Me6)RuCl2]2

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$174.00
1 G
$445.00

$174.00


Available to ship on16 April 2025Details


Request a Bulk Order

Select a Size

Change View
250 MG
$174.00
1 G
$445.00

About This Item

Empirical Formula (Hill Notation):
C24H36Cl4Ru2
CAS Number:
Molecular Weight:
668.49
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

$174.00


Available to ship on16 April 2025Details


Request a Bulk Order

form

solid

reaction suitability

core: ruthenium
reagent type: catalyst

mp

>300 °C

storage temp.

2-8°C

SMILES string

[Cl][Ru]Cl.[Cl][Ru]Cl.Cc1c(C)c(C)c(C)c(C)c1C.Cc2c(C)c(C)c(C)c(C)c2C

InChI

1S/2C12H18.4ClH.2Ru/c2*1-7-8(2)10(4)12(6)11(5)9(7)3;;;;;;/h2*1-6H3;4*1H;;/q;;;;;;2*+2/p-4

InChI key

GVCIEWAYDUQTTQ-UHFFFAOYSA-J

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service