Skip to Content
Merck
All Photos(1)

Key Documents

736309

Sigma-Aldrich

Poly(3,4-ethylenedioxythiophene), bis-poly(ethyleneglycol), lauryl terminated

0.8 wt. % (dispersion in 1,2-dichlorobenzene), contains p-toluenesulfonate as dopant

Synonym(s):

Aedotron P3-DCB, C12-PEG-block-PEDOT-block-PEG-C12, PEDOT:PEG

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352103
NACRES:
NA.23

form

liquid (dispersion)

contains

p-toluenesulfonate as dopant

composition

Aedotron TM-P3 polymer, 0.3-0.8 wt. %
1,2-Dichlorobenzene, 90-95 wt. %
ethanol, 2-8 wt. %
isopropanol, 0.2-0.8 wt. %
processing additive, 0.1-0.4 wt. %

concentration

0.6-1.0 wt. % (solid)
0.8 wt. % (dispersion in 1,2-dichlorobenzene)

refractive index

n20/D 1.532

conductivity

0.01-0.05 S/cm (bulk)

density

1.232 g/mL at 25 °C

Looking for similar products? Visit Product Comparison Guide

General description

Aedotron polymers are solvent-dispersable forms of PEDOT. These are copolymers of doped PEDOT and a flexible, soluble polymer such as poly(ethylene glycol) (PEG). These colloidal dispersions are stabilized by the highly solvated PEG chains which sterically limit the aggregation of the PEDOT blocks. Aedotron polymers are neither acidic nor corrosive, it easily disperses in polar aprotic solvents.
Sheet resistance, spin cast thin films: 106-108 Ohms/sq

Average transmittance: 60-85%

General purpose, moderately conductive dispersion in chlorinated solvents

Application

Conducting polymer dispersion suitable for making conducting films, anti-static layers and transparent electrodes. 1,2 Dichlorobenzene can be used to cast robust films on many polymeric substrates, but should be tested in each case.

Features and Benefits

The block copolymer structure renders the conducting polymer PEDOT highly dispersible in organic solvents. dichlorobenzene is compatible with many coatings and polymers formulations.

Other Notes

May settle over time. Must be sonicated before use

Legal Information

Aedotron is a trademark of TDA Research, Inc.

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

109.9 °F

Flash Point(C)

43.3 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Progress in Organic Thermoelectric Materials & Devices including high ZT values of >0.2 at room temperature by p-type (PEDOT:PSS) & n-type (Poly[Kx(Ni-ett)]) materials are discussed.

The application of conducting polymers at the interface with biology is an exciting new trend in organic electronics research.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service