Skip to Content
Merck
All Photos(1)

Key Documents

735000

Sigma-Aldrich

Arachidonic-5,6,8,9,11,12,14,15-d8 acid

≥98 atom % D, ≥98% (CP)

Synonym(s):

cis,cis,cis,cis-5,8,11,14-Eicosatetraenoic acid-5,6,8,9,11,12,14,15-d8

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$908.00

$908.00


Check Cart for Availability

Request more information

Select a Size

Change View
5 MG
$908.00

About This Item

Linear Formula:
CH3(CH2)4(CD=CDCH2)4CH2CH2CO2H
CAS Number:
Molecular Weight:
312.52
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.24

$908.00


Check Cart for Availability

Request more information

isotopic purity

≥98 atom % D

Quality Level

Assay

≥98% (CP)

form

liquid

bp

169-171 °C/0.15 mmHg (lit.)

mp

-49 °C (lit.)

mass shift

M+8

shipped in

dry ice

storage temp.

−20°C

SMILES string

[2H]\C(CCCCC)=C(/[2H])C\C([2H])=C(\[2H])C\C([2H])=C(\[2H])C\C([2H])=C(\[2H])CCCC(O)=O

InChI

1S/C20H32O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20(21)22/h6-7,9-10,12-13,15-16H,2-5,8,11,14,17-19H2,1H3,(H,21,22)/b7-6-,10-9-,13-12-,16-15-/i6D,7D,9D,10D,12D,13D,15D,16D

InChI key

YZXBAPSDXZZRGB-FBFLGLDDSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service