Skip to Content
Merck
All Photos(1)

Key Documents

73469

Sigma-Aldrich

(SP,S′P)-1,1′-Bis[bis(4-methoxy-3,5-dimethylphenyl)phosphino]-2,2′-bis[(R)-α-(dimethylamino)benzyl]ferrocene

≥97%

Synonym(s):

(1R,1′R)- 1,1′-Bis[bis(4-methoxy-3,5-dimethylphenyl)phosphino]-2,2′-bis[(R)-(dimethylamino)phenylmethyl]ferrocene (acc to CAS), Mandyphos SL-M004-1

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$126.00
1 G
$775.00
50 G
Inquire
100 G
Inquire

$126.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
$126.00
1 G
$775.00
50 G
Inquire
100 G
Inquire

About This Item

Empirical Formula (Hill Notation):
C64H74FeN2O4P2
CAS Number:
Molecular Weight:
1053.08
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

$126.00


Check Cart for Availability

Request a Bulk Order

Assay

≥97%

form

powder

optical purity

ee: ≥99%

functional group

amine
phenyl
phosphine

SMILES string

[Fe].COc1c(C)cc(cc1C)P([C]2[CH][CH][CH][C]2[C@H](N(C)C)c3ccccc3)c4cc(C)c(OC)c(C)c4.COc5c(C)cc(cc5C)P([C]6[CH][CH][CH][C]6[C@H](N(C)C)c7ccccc7)c8cc(C)c(OC)c(C)c8

InChI

1S/2C32H37NO2P.Fe/c2*1-21-17-26(18-22(2)31(21)34-7)36(27-19-23(3)32(35-8)24(4)20-27)29-16-12-15-28(29)30(33(5)6)25-13-10-9-11-14-25;/h2*9-20,30H,1-8H3;/t2*30-;/m11./s1

InChI key

HEJIDMKBSDYNOY-LJXMUBMNSA-N

General description

Mandyphos SL-M004-1 is a chiral ferrocenyl based diphosphine ligand belonging to the Mandyphos family.[1]

Application

It may be used as a chiral ligand in the rhodium complex-catalyzed hydrogenation reaction, a key step during the synthesis of argatroban.[2]

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

sold in collaboration with Solvias AG

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Diastereoselective synthesis of an argatroban intermediate, ethyl (2R, 4R)-4-methylpipecolate, by means of a Mandyphos/rhodium complex-catalyzed hydrogenation.
Ferraboschi P, et al.
Tetrahedron Asymmetry, 22(16), 1626-1631 (2011)
Modular chiral ligands: the profiling of the Mandyphos and Taniaphos ligand families.
Spindler F, et al.
Tetrahedron Asymmetry, 15(14), 2299-2306 (2004)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service