Skip to Content
Merck
All Photos(1)

Key Documents

734004

Sigma-Aldrich

4-(Trifluoromethyl)pyridazine

97%

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
$74.80

$74.80


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 MG
$74.80

About This Item

Empirical Formula (Hill Notation):
C5H3F3N2
CAS Number:
Molecular Weight:
148.09
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$74.80


Check Cart for Availability

Request a Bulk Order

Assay

97%

form

liquid

refractive index

n20/D 1.427

density

1.400 g/mL at 25 °C

functional group

fluoro

SMILES string

FC(F)(F)c1ccnnc1

InChI

1S/C5H3F3N2/c6-5(7,8)4-1-2-9-10-3-4/h1-3H

InChI key

RKYLAEZHYUWJRS-UHFFFAOYSA-N

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

187.0 °F

Flash Point(C)

86.11 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service