Skip to Content
Merck
All Photos(3)

Documents

733482

Sigma-Aldrich

Bismuth(III) telluride

greener alternative

powder, −325 mesh, 99.99% trace metals basis

Synonym(s):

Bismuth sesquitelluride, Bismuth telluride, Dibismuth tritelluride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Bi2Te3
CAS Number:
Molecular Weight:
800.76
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

Assay

99.99% trace metals basis

form

powder

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

particle size

−325 mesh

mp

573 °C
585 °C (lit.)

density

7.642 g/mL at 25 °C (lit.)

greener alternative category

SMILES string

[Te]=[Bi][Te][Bi]=[Te]

InChI

1S/2Bi.3Te

InChI key

GUYIRKJSQUOSJV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Bismuth(III) telluride (Bi2Te3) is a thermoelectric material, which acts as a topological insulator: a quantum material with an insulating bulky gap and gapless edge. It forms thin films by different techniques, which include chemical method, electrodeposition and molecular jet method.
The binary chalcogenide, bismuth(III) telluride occurs as mineral tellurobismuthite in nature. 1-2
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Application

Bi2Te3 can be used for a saturable absorber in a variety of photo-optic applications, which include pulse fiber lasers.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Organic-inorganic nanohybrids as novel thermoelectric materials: hybrids of polyaniline and bismuth (III) telluride nanoparticles
Toshima N, et al.
Journal of Electronic Materials, 40(5), 898-902 (2011)
IOP Conference Series: Materials Science and Engineering, 210(1), 012032-012032 (2017)
Journal of Physics. Conference Series, 1151(1), 012002-012002 (2019)

Articles

Thermoelectric materials comprise a wide range of solid compounds distinguished by their ability to convert thermal and electrical energy.

Professors summarize recent 2D materials synthesis advancements and biosensing applications in various fields.

Higher transition metal silicides are ideal for anisotropic thermoelectric conversion due to their Seebeck coefficient anisotropy and mechanical properties.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service