Skip to Content
Merck
All Photos(1)

Key Documents

733342

Sigma-Aldrich

9-Fluorenyl methacrylate

97%

Synonym(s):

9-FMA, FMA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H14O2
CAS Number:
Molecular Weight:
250.29
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

form

solid

mp

60-65 °C

storage temp.

2-8°C

SMILES string

CC(=C)C(=O)OC1c2ccccc2-c3ccccc13

InChI

1S/C17H14O2/c1-11(2)17(18)19-16-14-9-5-3-7-12(14)13-8-4-6-10-15(13)16/h3-10,16H,1H2,2H3

InChI key

IFAFXDPXPSZLKZ-UHFFFAOYSA-N

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Haneul Kim et al.
Scientific reports, 7, 41090-41090 (2017-01-21)
Targeted drug delivery has long been extensively researched since drug delivery and release at the diseased site with minimum dosage realizes the effective therapy without adverse side effects. In this work, to achieve enhanced intracellular uptake of anticancer drug carriers
Christopher Ochs et al.
Journal of biomedical informatics, 71, 165-177 (2017-06-07)
Biomedical ontologies often reuse content (i.e., classes and properties) from other ontologies. Content reuse enables a consistent representation of a domain and reusing content can save an ontology author significant time and effort. Prior studies have investigated the existence of
Pierre Nicolo et al.
Archives of physical medicine and rehabilitation, 99(5), 862-872 (2017-12-11)
To investigate the effects of cathodal transcranial direct current stimulation (tDCS) and continuous theta burst stimulation (cTBS) on neural network connectivity and motor recovery in individuals with subacute stroke. Double-blinded, randomized, placebo-controlled study. University hospital rehabilitation unit. Inpatients with stroke
Gary W Mack
Journal of applied physiology (Bethesda, Md. : 1985), 123(2), 317-325 (2017-05-20)
Quantitative assessment of small-fiber peripheral neuropathy often involves an evaluation of the interaction between the C-fiber sudomotor nerve and local sweat rate (SR). Typically, some sort of quantitative sudomotor axon reflex test (QSART) is performed to aid in diagnosing small-fiber
Seunghee Lee et al.
IEEE transactions on neural systems and rehabilitation engineering : a publication of the IEEE Engineering in Medicine and Biology Society, 26(1), 125-134 (2017-09-28)
The Fugl-Meyer assessment (FMA) is the most popular instrument for evaluating upper extremity motor function in stroke patients. However, it is a labor-intensive and time-consuming method. This paper proposes a novel automated FMA system to overcome these limitations of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service