Skip to Content
Merck
All Photos(1)

Key Documents

732818

Sigma-Aldrich

7-Bromochromone

Synonym(s):

7-Bromo-4H-chromen-4-one

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
CHF 229.07
100 MG
CHF 343.56
250 MG
CHF 521.94

Vendor SKU: W11413-50MG

CHF 229.07


Ships in 14 days - from Aldrich Partner.

Request a Bulk Order

Select a Size

Change View
50 MG
CHF 229.07
100 MG
CHF 343.56
250 MG
CHF 521.94

About This Item

Empirical Formula (Hill Notation):
C9H5BrO2
CAS Number:
Molecular Weight:
225.04
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Vendor SKU: W11413-50MG

CHF 229.07


Ships in 14 days - from Aldrich Partner.

Request a Bulk Order

form

solid

mp

154-159 °C

SMILES string

Brc1ccc2C(=O)C=COc2c1

InChI

1S/C9H5BrO2/c10-6-1-2-7-8(11)3-4-12-9(7)5-6/h1-5H

InChI key

WWAOCYNUAWVKGQ-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service