Skip to Content
Merck
All Photos(2)

Key Documents

732435

Sigma-Aldrich

4,7-Bis(2-bromo-5-thienyl)-2,1,3-benzothiadiazole

≥99.0% (HPLC)

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$624.00

$624.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$624.00

About This Item

Empirical Formula (Hill Notation):
C14H6Br2N2S3
CAS Number:
Molecular Weight:
458.21
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

$624.00


Check Cart for Availability

Request a Bulk Order

Assay

≥99.0% (HPLC)

form

solid

mp

245-247 °C

SMILES string

Brc1ccc(s1)-c2ccc(-c3ccc(Br)s3)c4nsnc24

InChI

1S/C14H6Br2N2S3/c15-11-5-3-9(19-11)7-1-2-8(10-4-6-12(16)20-10)14-13(7)17-21-18-14/h1-6H

InChI key

ZIIMIGRZSUYQGW-UHFFFAOYSA-N

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bryce McCarthy et al.
Scientific reports, 11(1), 1532-1532 (2021-01-17)
Colorectal cancer (CRC) treatment is currently hindered by micrometastatic relapse that cannot be removed completely during surgery and is often chemotherapy resistant. Targeted theranostic nanoparticles (NPs) that can produce heat for ablation and enable tumor visualization via their fluorescence offer

Articles

Organic electronics promise renewable energy solutions surpassing silicon-based tech.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service