Skip to Content
Merck
All Photos(1)

Key Documents

732192

Sigma-Aldrich

(1S,2S)-N-p-Tosyl-1,2-dimesitylethylenediamine

96%

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$196.00

$196.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
$196.00

About This Item

Empirical Formula (Hill Notation):
C27H34N2O2S
CAS Number:
Molecular Weight:
450.64
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

$196.00


Check Cart for Availability

Request a Bulk Order

Assay

96%

form

solid

mp

169-174 °C

functional group

amine

SMILES string

Cc1ccc(cc1)S(=O)(=O)N[C@H]([C@@H](N)c2c(C)cc(C)cc2C)c3c(C)cc(C)cc3C

InChI

1S/C27H34N2O2S/c1-16-8-10-23(11-9-16)32(30,31)29-27(25-21(6)14-18(3)15-22(25)7)26(28)24-19(4)12-17(2)13-20(24)5/h8-15,26-27,29H,28H2,1-7H3/t26-,27-/m0/s1

InChI key

XAXZXGHDWDUHFH-SVBPBHIXSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

The Chin group is interested in computational and experimental approaches to understanding stereoselective recognition and catalysis. Their studies in weak forces (H-bonding, electronic and steric effects) has led to a highly efficient method for making limitless varieties of chiral vicinal diamines from the 'mother diamine' that are useful for developing stereoselective organocatalysts or transition metal-based catalysts as well as for developing drugs (Acc Chem Res (2012) p1345). The 'mother diamine' is also useful for making binol, monophos and binap analogs. The Chin group is also interested in using reversible covalent bonds for stereoselective recognition and L to D conversion of natural and non-natural amino acids (EJOC (2012) p229).

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service