Skip to Content
Merck
All Photos(1)

Key Documents

730130

Sigma-Aldrich

Methyl 2-(trifluoromethyl)acrylate

contains <50 ppm 4-Hydroxy-TEMPO as stabilizer, 97%

Synonym(s):

MTFMA

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$683.00

$683.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$683.00

About This Item

Empirical Formula (Hill Notation):
C5H5F3O2
CAS Number:
Molecular Weight:
154.09
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

$683.00


Check Cart for Availability

Request a Bulk Order

Assay

97%

form

liquid

contains

<50 ppm 4-Hydroxy-TEMPO as stabilizer

refractive index

n20/D 1.359

density

1.262 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

COC(=O)C(=C)C(F)(F)F

InChI

1S/C5H5F3O2/c1-3(4(9)10-2)5(6,7)8/h1H2,2H3

InChI key

GTRBXMICTQNNIN-UHFFFAOYSA-N

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

73.0 °F - closed cup

Flash Point(C)

22.8 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service