Skip to Content
Merck
All Photos(1)

Documents

729493

Sigma-Aldrich

2,4-Dichloro-5-fluoropyrimidine-2-13C,15N2

99 atom % 13C, 98 atom % 15N, 98% (CP)

Synonym(s):

2,4-dichloro-5-fluoro-Pyrimidine-2-13C-1,3-15N2, Uridine-13C9-15N2 5′-(tetrahydrogen triphosphate), 2′-deoxy-, sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13CC3HCl2F15N2
CAS Number:
Molecular Weight:
169.95
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

isotopic purity

99 atom % 13C
98 atom % 15N

Assay

98% (CP)

form

solid

mp

37-41 °C (lit.)

application(s)

agriculture

mass shift

M+3

SMILES string

Fc1c[15n][13c](Cl)[15n]c1Cl

InChI

1S/C4HCl2FN2/c5-3-2(7)1-8-4(6)9-3/h1H/i4+1,8+1,9+1

InChI key

WHPFEQUEHBULBW-WKPWXFLVSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Corr. 1B

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

222.8 °F - closed cup

Flash Point(C)

106 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Tracer techniques using isotopic N have been employed since the 1940s to investigate such topics as N transformations and cycling in agricultural and other soils, N sources for crop uptake, the fate and behavior of fertilizer N applied to soils, the nature and extent of soil and fertilizer losses, and fertilizer N uptake efficiency.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service