Skip to Content
Merck
All Photos(1)

Documents

729183

Sigma-Aldrich

Lanthanum nickelate, strontium doped

greener alternative

nanopowder, <100 nm particle size (BET)

Synonym(s):

LaSNO, Lanthanum nickel strontium oxide

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
26111700
NACRES:
NA.23

form

nanopowder

mol wt

Mw 379.99 g/mol

composition

La1.6Sr0.4NiO4

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

surface area

5-10 m2/g

particle size

<100 nm (BET)

mp

>400 °C

bulk density

0.818 g/mL

application(s)

battery manufacturing

greener alternative category

General description

Strontium-doped lanthanum nickelate can be prepared by combined precipitation and molten salt reactions at 1073K.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Click here for more details.

Application

Lanthanum nickelate, strontium doped can be used in the fabrication of an electrode material with a specific capacitance value of 719 F g−1 at a current density of 2 A g−1. It can further be used in the formation of high energy density supercapacitors.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Crespin, M; Bassat, J. M.; Odier, P.; Mouron, P.; Choisnet, J.
Journal of Solid State Chemistry, 84, 165-165 (1990)
Preparation and characterisation of lanthanum nickel strontium oxides by combined coprecipitation and molten salt reactions
Kao CF, et al.
Ceramics International, 26(3), 237-243 (2000)
Gong, M; Lu, L.; Zhang, H.; Gao, L.; Guo, Y.; Jin, J.
Mrs Bulletin, 1630-1630 (2009)
Sr-doped lanthanum nickelate nanofibers for high energy density supercapacitors
Cao Y, et al.
Electrochimica Acta, 174, 41-50 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service