Skip to Content
Merck
All Photos(1)

Key Documents

72915

Sigma-Aldrich

Sodium 3-nitrobenzoate

≥95.0% (HPLC)

Synonym(s):

3-Nitrobenzoic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
3-O2NC6H4CO2Na
CAS Number:
Molecular Weight:
189.10
Beilstein:
4166479
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥95.0% (HPLC)

form

solid

SMILES string

[Na+].[O-]C(=O)c1cccc(c1)[N+]([O-])=O

InChI

1S/C7H5NO4.Na/c9-7(10)5-2-1-3-6(4-5)8(11)12;/h1-4H,(H,9,10);/q;+1/p-1

InChI key

MUADFEZFSKAZLT-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Sodium 3-nitrobenzoate may be used in the synthesis of [Ni(cyclam)(3-nitrobenzoate)2] complex. [Cyclam = 1,4,8,11-tetraazacyclotetradecane][1]

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Sol. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Novel nickel cyclam complexes with potent antimicrobial and cytotoxic properties.
Al-Bari MAA
Journal of Applied Sciences Research, 3(11), 1251-1261 (2007)
Zhiqiang Hu et al.
Water research, 39(4), 710-720 (2005-02-15)
The physical and biochemical characteristics of the biomass in three lab-scale sequencing batch reactors (SBR) treating a synthetic wastewater at a 20-day target solids retention time (SRT) were investigated. The synthetic wastewater feed contained biogenic compounds and 22 organic priming
Reproductive toxicology. m-Nitrobenzoic acid.
Environmental health perspectives, 105 Suppl 1, 325-326 (1997-02-01)
Miguel A Providenti et al.
Applied and environmental microbiology, 72(4), 2651-2660 (2006-04-07)
In Comamonas sp. strain JS46, 3-nitrobenzoate (3Nba) is initially oxidized at the 3,4 position by a dioxygenase, which results in release of nitrite and production of protocatechuate. The locus coding for the 3Nba dioxygenase (designated mnb, for m-nitrobenzoate) was mobilized
M Samsonowicz et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 82(1), 235-246 (2011-08-25)
The influence of beryllium, magnesium, calcium, strontium and barium cations on the electronic system of 3-nitrobenzoic acid was studied in comparison with studied earlier alkali metal ions. The vibrational FT-IR (in KBr and ATR techniques) and (1)H and (13)C NMR

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service