Skip to Content
Merck
All Photos(1)

Key Documents

728942

Sigma-Aldrich

3-Bromo-2-fluorotoluene

97%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$96.40

$96.40


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$96.40

About This Item

Empirical Formula (Hill Notation):
C7H6BrF
CAS Number:
Molecular Weight:
189.02
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$96.40


Check Cart for Availability

Request a Bulk Order

Assay

97%

form

liquid

refractive index

n20/D 1.533

density

1.503 g/mL at 25 °C

functional group

bromo
fluoro

SMILES string

Cc1cccc(Br)c1F

InChI

1S/C7H6BrF/c1-5-3-2-4-6(8)7(5)9/h2-4H,1H3

InChI key

LZVNGSFAHGKCDM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

170.1 °F

Flash Point(C)

76.7 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service