Skip to Content
Merck
All Photos(2)

Key Documents

728543

Sigma-Aldrich

(S)-(−)-α,α-Diphenyl-2-pyrrolidinemethanol tert-butyldimethylsilyl ether

≥97% (HPLC)

Synonym(s):

(S)-(−)-α,α-Diphenylprolinol tert-butyldimethylsilyl ether, (S)-2-[(tert-Butyldimethylsiloxy)diphenylmethyl]pyrrolidine

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$890.00

$890.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$890.00

About This Item

Empirical Formula (Hill Notation):
C23H33NOSi
CAS Number:
Molecular Weight:
367.60
Beilstein:
10368174
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

$890.00


Check Cart for Availability

Request a Bulk Order

Assay

≥97% (HPLC)

form

powder

optical purity

enantiomeric excess: ≥98.0%

functional group

phenyl

SMILES string

CC(C)(C)[Si](C)(C)OC([C@@H]1CCCN1)(c2ccccc2)c3ccccc3

InChI

1S/C23H33NOSi/c1-22(2,3)26(4,5)25-23(21-17-12-18-24-21,19-13-8-6-9-14-19)20-15-10-7-11-16-20/h6-11,13-16,21,24H,12,17-18H2,1-5H3/t21-/m0/s1

InChI key

YJFSFDYMOMREQP-NRFANRHFSA-N

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service