Skip to Content
Merck
All Photos(1)

Key Documents

726672

Sigma-Aldrich

(S)-(+)-2-Butanol

ChiPros®, produced by BASF, 99%

Synonym(s):

(S)-(+)-sec-Butyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5CH(OH)CH3
CAS Number:
Molecular Weight:
74.12
Beilstein:
1718763
EC Number:
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:

grade

produced by BASF

vapor density

2.6 (vs air)

vapor pressure

12.5 mmHg ( 20 °C)

Assay

≥98.5%
99%

form

liquid

optical purity

enantiomeric excess: ≥98.5%

expl. lim.

9.8 %

refractive index

n20/D 1.397 (lit.)

bp

99-100 °C (lit.)

density

0.803 g/mL at 25 °C (lit.)

SMILES string

CC[C@H](C)O

InChI

1S/C4H10O/c1-3-4(2)5/h4-5H,3H2,1-2H3/t4-/m0/s1

InChI key

BTANRVKWQNVYAZ-BYPYZUCNSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

ChiPros is a registered trademark of BASF SE

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

75.2 °F - closed cup

Flash Point(C)

24 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaofeng Hu et al.
The journal of physical chemistry. A, 115(23), 5785-5793 (2011-01-26)
The adsorption, thermal evolution, and electron irradiation of 2-butanol on Pt(111) were investigated with reflection absorption infrared spectroscopy (RAIRS). A simulated vibrational spectrum of a single 2-butanol molecule was calculated using density functional theory to facilitate vibrational assignments. Exposures of
Prabha Dwivedi et al.
Analytical chemistry, 78(24), 8200-8206 (2006-12-15)
This article introduces the concept of chiral ion mobility spectrometry (CIMS) and presents examples demonstrating the gas-phase separation of enantiomers of a wide range of racemates including pharmaceuticals, amino acids, and carbohydrates. CIMS is similar to traditional ion mobility spectrometry
Kari V Ahonen et al.
Steroids, 77(11), 1141-1151 (2012-07-21)
Synthesis and solid-state structural characterization of five bile acid amides of 4-aminopyridine (4-AP) are reported. Systematic crystallization experiments revealed a number of structural modifications and/or solvate/hydrate systems for these conjugates. Particularly, cholic acid conjugate exhibited five distinct structure modifications, including
Patrycja Seroka et al.
Magnetic resonance in chemistry : MRC, 44(2), 132-138 (2005-12-17)
D-Glucose, D-mannose and L-rhamnose were reacted with a racemic mixture of 2-butanol, and the resulting alpha-glycosides were analyzed by 1H NMR with COSY and NOESY experiments. Conformational analysis of alpha-glycosidic bonds performed with molecular modeling and appropriate heteronuclear long-range coupling
Charles O Stanier et al.
Environmental science & technology, 41(8), 2756-2763 (2007-05-31)
The temperature-dependence of secondary organic aerosol (SOA) concentrations is measured using a temperature-controlled smog chamber. Aerosols are generated from reaction of alpha-pinene (14-150 ppb) and ozone at a constant temperature of 22 +/- 2 degrees C in the presence of

Articles

Chiral alcohols form a versatile class of chiral synthons, since they can be incorporated into the API structures directly as esters or ethers.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service