Skip to Content
Merck
All Photos(1)

Key Documents

726370

Furfuryl chloride solution

Synonym(s):

2-(Chloromethyl)furan solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H5ClO
CAS Number:
Molecular Weight:
116.55
Beilstein:
106288
UNSPSC Code:
12352101

InChI

1S/C5H5ClO/c6-4-5-2-1-3-7-5/h1-3H,4H2

InChI key

GANBJDIOIDQSGI-UHFFFAOYSA-N


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service