Skip to Content
Merck
All Photos(2)

Key Documents

725544

Sigma-Aldrich

Tetrakis(ethylmethylamido)hafnium(IV)

packaged for use in deposition systems

Synonym(s):

TEMAH, Tetrakis(ethylmethylamino)hafnium(IV)

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$1,810.00

$1,810.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 G
$1,810.00

About This Item

Linear Formula:
[(CH3)(C2H5)N]4Hf
CAS Number:
Molecular Weight:
410.90
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

$1,810.00


Check Cart for Availability

Request a Bulk Order

Quality Level

form

liquid

reaction suitability

core: hafnium
reagent type: catalyst

bp

78 °C/0.01 mmHg (lit.)

mp

<-50 °C

density

1.324 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CCN(C)[Hf](N(C)CC)(N(C)CC)N(C)CC

InChI

1S/4C3H8N.Hf/c4*1-3-4-2;/h4*3H2,1-2H3;/q4*-1;+4

InChI key

NPEOKFBCHNGLJD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Tetrakis(ethylmethylamido)hafnium(IV) (TEMAH) is a colorlessliquid, which freezes at -50 °C and boils around 78 °C at 0.1 Torr. It is air-and water-sensitive.

Application

TEMAH is used as a precursor for atomic layer deposition(ALD) of conformal thin films of hafnium oxide (HfO2) and hafnium zirconium oxide (Hf1-xZrxO2), [1] which are used as dielectric films in semiconductor fabrication because of their high dielectric constants.

TEMAH is well-suited for ALD because its adsorption is self-limiting on a number of substrates including glass, indium-tin oxide(ITO), Si(100), and two-dimensional materials like MoS2.[2][3] TEMAH also conveniently reacts with either water or ozone as the oxygen-source in the ALD process.

Features and Benefits

This TEMAH is packaged in a Swagelok stainless-steeldeposition system convenient for connecting to ALD systems.

  • Steel cylinder connected to 316 stainless steelball-valve
  • 1/4 inch male Swagelok VCR connections

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B - STOT SE 3 - Water-react 1

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Atomic Layer Deposition of Hafnium Dioxide Films from Hafnium Tetrakis(ethylmethylamide) and Water.
Kukli K, et al.
Chem. Vap. Deposition, 199-204 (2002)
Martin Rose et al.
ACS applied materials & interfaces, 2(2), 347-350 (2010-04-02)
The mechanisms of technologically important atomic layer deposition (ALD) processes, trimethylaluminium (TMA)/ozone and tetrakis(ethylmethylamino)hafnium (TEMAH)/ozone, for the growth of Al(2)O(3) and HfO(2) thin films are studied in situ by a quadrupole mass spectrometer coupled with a 300 mm ALD reactor.
Jaehyun Yang et al.
ACS applied materials & interfaces, 5(11), 4739-4744 (2013-05-21)
We report on the effect of oxygen plasma treatment of two-dimensional multilayer MoS2 crystals on the subsequent growth of Al2O3 and HfO2 films, which were formed by atomic layer deposition (ALD) using trimethylaluminum and tetrakis-(ethylmethylamino)hafnium metal precursors, respectively, with water
Luqi Tu et al.
Nature communications, 11(1), 101-101 (2020-01-05)
Sensitive photodetection is crucial for modern optoelectronic technology. Two-dimensional molybdenum disulfide (MoS2) with unique crystal structure, and extraordinary electrical and optical properties is a promising candidate for ultrasensitive photodetection. Previously reported methods to improve the performance of MoS2 photodetectors have focused

Articles

Atomic Layer Deposition (ALD) technology ensures uniform coating on complex 3D surfaces with precise chemisorption cycles.

Nanocomposite Coatings with Tunable Properties Prepared by Atomic Layer Deposition

Thin film photovoltaic devices have become increasingly important in efficiently harnessing solar energy to meet consumer demand.

The properties of many devices are limited by the intrinsic properties of the materials that compose them.

Questions

  1. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service