Skip to Content
Merck
All Photos(3)

Key Documents

725471

Sigma-Aldrich

Bis(methyl-η5−cyclopentadienyl)methoxymethylzirconium

packaged for use in deposition systems

Synonym(s):

ZRCMMM, ZrD-CO4

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$1,590.00

$1,590.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 G
$1,590.00

About This Item

Linear Formula:
Zr(CH3C5H4)2CH3OCH3
Molecular Weight:
295.53
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

$1,590.00


Check Cart for Availability

Request a Bulk Order

form

liquid

reaction suitability

core: zirconium

color

colorless

bp

110 °C/0.5 mmHg (lit.)

density

1.27 g/mL±0.01 g/mL at 25 °C (lit.)

SMILES string

C[C]1[C][C][C][C]1.C[C]2[C][C][C][C]2.C[Zr]OC

InChI

1S/2C6H7.CH3O.CH3.Zr/c2*1-6-4-2-3-5-6;1-2;;/h2*2-5H,1H3;1H3;1H3;/q;;-1;;+1

InChI key

LFGIFPGCOXPKMG-UHFFFAOYSA-N

General description

Atomic number of base material: 40 Zirconium

Application

Advanced precursor for atomic layer deposition of ZrO2 thin films. Hafnium and zirconium oxides are leading candidates to replace silicion dioxide as the gate oxide in a variety of semiconductor and energy applications.[1] Excellent properties of HfO2 and ZrO2 films make them especially attractive for gate oxide replacement and as potential insulating dielectrics for capacitive elements in memory devices such as DRAM.[2]

Features and Benefits

Compatible with a variety of oxidants in ALD growth processes across a wide temperature range exhibiting self limiting growth up to 400 °C. Precursor volatility and thermal stability properties enable easy materials transport from bubblers into conventional deposition tools.

Packaging

Packaged in stainless steel cylinders compatible with conventional deposition systems. Precursors may be used in liquid injection systems as dilute solutions and in combination with a variety of other sources to deposit mixed oxides.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids not in Storage Class 3

WGK

WGK 3

Flash Point(F)

226.4 °F

Flash Point(C)

108 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

J. W. Elama
Applied Physics Letters, 91, 253123-253123 (2007)
Gutsche, M.; Seidl, H.; Luetzen J.; Birner, A.;
International Electron Devices Meeting, 18-18 (2001)
High-k gate dielectrics: current status and material properties considerations
Wilk, G.D.; Wallace, R.M.; Anthony, J.M.
Journal of Applied Physics, 89, 5243-5243 (2001)

Articles

Atomic Layer Deposition (ALD) technology ensures uniform coating on complex 3D surfaces with precise chemisorption cycles.

Nanocomposite Coatings with Tunable Properties Prepared by Atomic Layer Deposition

Thin film photovoltaic devices have become increasingly important in efficiently harnessing solar energy to meet consumer demand.

Nanomaterials are considered a route to the innovations required for large-scale implementation of renewable energy technologies in society to make our life sustainable.

See All

Questions

  1. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service