Skip to Content
Merck
All Photos(1)

Key Documents

725374

Sigma-Aldrich

2,2-Dibutyl-[1,3,2]dioxastannolane

96%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H22O2Sn
CAS Number:
Molecular Weight:
292.99
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

form

solid

mp

231-236 °C

SMILES string

CCCC[Sn]1(CCCC)OCCO1

InChI

1S/2C4H9.C2H4O2.Sn/c2*1-3-4-2;3-1-2-4;/h2*1,3-4H2,2H3;1-2H2;/q;;-2;+2

InChI key

STWBEJNHFFEKHC-UHFFFAOYSA-N

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service