Skip to Content
Merck
All Photos(2)

Key Documents

724793

Sigma-Aldrich

3-Nitropyridine-2-thiol

96%

Synonym(s):

3-Nitro-2-pyridinethiol

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$158.00

$158.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$158.00

About This Item

Empirical Formula (Hill Notation):
C5H4N2O2S
CAS Number:
Molecular Weight:
156.16
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$158.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

96%

form

solid

mp

169-175 °C

functional group

nitro

SMILES string

[O-][N+](=O)c1cccnc1S

InChI

1S/C5H4N2O2S/c8-7(9)4-2-1-3-6-5(4)10/h1-3H,(H,6,10)

InChI key

LKNPLDRVWHXGKZ-UHFFFAOYSA-N

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H Zhang et al.
Thrombosis and haemostasis, 112(6), 1304-1311 (2014-09-19)
Dual antiplatelet therapy with clopidogrel and aspirin has been the standard of care in the United States for patients with acute coronary syndromes (ACS) and/or undergoing percutaneous coronary interventions (PCI). However, the effectiveness of clopidogrel varies significantly among different sub-populations

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service