Skip to Content
Merck
All Photos(1)

Key Documents

723789

Sigma-Aldrich

5-Methoxy-6-(trifluoromethyl)indole

97%

Synonym(s):

5-Methoxy-6-trifluoromethyl-1H-indole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H8F3NO
CAS Number:
Molecular Weight:
215.17
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

form

solid

mp

104-105 °C

functional group

fluoro

SMILES string

COc1cc2cc[nH]c2cc1C(F)(F)F

InChI

1S/C10H8F3NO/c1-15-9-4-6-2-3-14-8(6)5-7(9)10(11,12)13/h2-5,14H,1H3

InChI key

KNMAZUXTEPXPOM-UHFFFAOYSA-N

Application

  • Reactant for preparation of pyridylcarbamoylindolines as selective 5HT2C/2B receptor antagonists

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service