Skip to Content
Merck
All Photos(2)

Key Documents

723339

Sigma-Aldrich

2-Bromo-4-fluoropyridine

96%

Synonym(s):

4-Fluoro-2-bromopyridine

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$106.00

$106.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$106.00

About This Item

Empirical Formula (Hill Notation):
C5H3BrFN
CAS Number:
Molecular Weight:
175.99
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$106.00


Check Cart for Availability

Request a Bulk Order

Assay

96%

form

liquid

refractive index

n20/D 1.537

density

1.699 g/mL at 25 °C

functional group

bromo
fluoro

SMILES string

Fc1ccnc(Br)c1

InChI

1S/C5H3BrFN/c6-5-3-4(7)1-2-8-5/h1-3H

InChI key

AIEATTRZKVGMBO-UHFFFAOYSA-N

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

170.1 °F - closed cup

Flash Point(C)

76.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service