Skip to Content
Merck
All Photos(2)

Documents

723134

Sigma-Aldrich

Tris(2-pyridylmethyl)amine

98%

Synonym(s):

TPA, TPMA, Tri-(2-picolyl)amine, Tripicolylamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H18N4
CAS Number:
Molecular Weight:
290.36
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

98%

form

solid

mp

85-89 °C

SMILES string

C(N(Cc1ccccn1)Cc2ccccn2)c3ccccn3

InChI

1S/C18H18N4/c1-4-10-19-16(7-1)13-22(14-17-8-2-5-11-20-17)15-18-9-3-6-12-21-18/h1-12H,13-15H2

InChI key

VGUWFGWZSVLROP-UHFFFAOYSA-N

Application

Atom Transfer Radical Polymerization (ATRP) ligand for the creation of telechelic polymers.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tsarevsky, N. V.; Braunecker, W. A.; Brooks, S. J.; Matyjaszewski, K.
Macromolecules, 39, 6817-6817 (2006)
Christiane Trimpert et al.
PloS one, 12(9), e0183774-e0183774 (2017-09-21)
Regulation of our water homeostasis is fine-tuned by dynamic translocation of Aquaporin-2 (AQP2)-bearing vesicles to and from the plasma membrane of renal principal cells. Whereas binding of vasopressin to its type-2 receptor initiates a cAMP-protein kinase A cascade and AQP2
Po-Hsiung Kung et al.
Redox biology, 13, 266-277 (2017-06-11)
Protein disulfide isomerase (PDI) present at platelet surfaces has been considered to play an important role in the conformational change and activation of the integrin glycoprotein IIb/IIIa (GPIIb/IIIa) and thus enhances platelet aggregation. Growing evidences indicated that platelet surface PDI
Xia, J.; Gaynor, S. G.; Matyjaszewski, K.
Macromolecules, 31, 5958-5958 (1998)
Andrea Bilger et al.
Oncotarget, 8(27), 44266-44280 (2017-06-03)
EBV infection causes mononucleosis and is associated with specific subsets of B cell lymphomas. Immunosuppressed patients such as organ transplant recipients are particularly susceptible to EBV-induced lymphoproliferative disease (LPD), which can be fatal. Leflunomide (a drug used to treat rheumatoid

Articles

ATRP is a successful method for precise polymer synthesis with controlled molecular weights and high chain end functionalities.

Micro review of reversible addition/fragmentation chain transfer (RAFT) polymerization.

Protocols

We present an article about RAFT, or Reversible Addition/Fragmentation Chain Transfer, which is a form of living radical polymerization.

We presents an article featuring procedures that describe polymerization of methyl methacrylate and vinyl acetate homopolymers and a block copolymer as performed by researchers at CSIRO.

Polymerization via ATRP procedures demonstrated by Prof. Dave Haddleton's research group at the University of Warwick.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service