Skip to Content
Merck
All Photos(3)

Key Documents

722251

Sigma-Aldrich

7-Fluoro-1-tetralone

97%

Synonym(s):

7-Fluoro-3,4-dihydronaphthalen-1(2H)-one

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$98.50
5 G
$330.00

$98.50


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$98.50
5 G
$330.00

About This Item

Empirical Formula (Hill Notation):
C10H9FO
CAS Number:
Molecular Weight:
164.18
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$98.50


Check Cart for Availability

Request a Bulk Order

Assay

97%

form

solid

mp

61-66 °C

functional group

fluoro
ketone

storage temp.

2-8°C

SMILES string

Fc1ccc2CCCC(=O)c2c1

InChI

1S/C10H9FO/c11-8-5-4-7-2-1-3-10(12)9(7)6-8/h4-6H,1-3H2

InChI key

UCBYBFAJSWCTLG-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service