Skip to Content
Merck
All Photos(1)

Key Documents

722014

Sigma-Aldrich

6-Methoxyindole-2-carboxylic acid

95%

Synonym(s):

NSC 27988

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H9NO3
CAS Number:
Molecular Weight:
191.18
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

solid

mp

198-203 °C

SMILES string

COc1ccc2cc([nH]c2c1)C(O)=O

InChI

1S/C10H9NO3/c1-14-7-3-2-6-4-9(10(12)13)11-8(6)5-7/h2-5,11H,1H3,(H,12,13)

InChI key

XNBGANWAZJWOHS-UHFFFAOYSA-N

Application

  • Reactant for demethylation reactions using ionic liquids under microwave irradiation
  • Reactant for preparation of pyrazinoindoledione via Ugi reaction and microwave-assisted cyclization
  • Reactant for preparation of isoquinolinecarboxamides and their derivatives as opioid receptor antagonists
  • Synthetic intermediate for preparation of indole fatty alcohol derivatives
  • Reactant for preparation of acylaminoalkylindoles as MT2-selective melatonin antagonists

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service