Skip to Content
Merck
All Photos(1)

Key Documents

721921

Supelco

Methanol-d4

99.8 atom % D, NMR tube size 3 mm × 8 in.

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CD3OD
Molecular Weight:
36.07
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.12

isotopic purity

99.8 atom % D

Assay

99% (CP)

form

liquid

technique(s)

NMR: suitable

NMR tube size

3 mm × 8 in.

suitability

suitable for NMR (reference standard)

mass shift

M+4

Looking for similar products? Visit Product Comparison Guide

General description

Methanol-d4 (CD3OD) is a deuterated NMR solvent used for NMR-based research and analyses. It has the highest isotopic enrichment and will not interfere with the desired level of detection.
The absolute infrared absorption intensities of CD3OD have been reported between 8000 and 350cm-1.

Application

Methanol-d4 may be used as a removable internal standard to limit contamination during the quantification of lipophilic shellfish toxins using quantitative nuclear magnetic resonance technique (qNMR). The purity and ability of the lipophilic shellfish toxins to serve as authentic reference standards is also determined. It may also be used as a solvent in investigating the utility of matrix-assisted laser desorption ionization time-of-flight (MALDI-TOF) mass spectrometry to monitor peptide and protein conformational changes of bradykinin, α-melanocyte stimulating hormone, and melittin.

Quantity

3 mm O.D. tube contains 0.235 mL.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Matrix-assisted laser desorption ionization hydrogen/deuterium exchange studies to probe peptide conformational changes
Figueroa DI and Russell HD
Journal of the American Society For Mass Spectrometry, 10(8), 719-731 (1999)
Absolute quantification of lipophilic shellfish toxins by quantitative nuclear magnetic resonance using removable internal reference substance with SI traceability
Kato T, et al.
Analytical Sciences, 32(7), 729-734 (2016)
Infrared intensities of liquids. XVII. Infrared refractive indices from 8000 to 350 cm-1, absolute integrated absorption intensities, transition moments, and dipole moment derivatives of methan-d3-ol and methanol-d4 at 25?C.
Bertie JE and Zhang SL.
J. Chem. Phys. , 101(10), 8364-8379 (1994)
Cheng-Kun Lin et al.
Organic & biomolecular chemistry, 13(7), 2100-2107 (2014-12-20)
A straightforward synthesis of novel, 2-heterocyclyl polyhydroxylated pyrrolidines is described. Stereocontrolled additions of nucleophiles to cyclic nitrones generated the corresponding 2,3-trans adducts, allowing the synthesis of the corresponding pyrrolidines via key intermediates bearing an alkyne and a nitrile oxide. Three
Jon Kapla et al.
Physical chemistry chemical physics : PCCP, 17(34), 22438-22447 (2015-08-08)
The disaccharide trehalose (TRH) strongly affects the physical properties of lipid bilayers. We investigate interactions between lipid membranes formed by 1,2-dimyristoyl-sn-glycero-3-phosphocholine (DMPC) and TRH using NMR spectroscopy and molecular dynamics (MD) computer simulations. We compare dipolar couplings derived from DMPC/TRH

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service