721387
3-(2-Hydroxy-2-propanyl)phenylboronic acid pinacol ester
97%
Synonym(s):
2-(3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)propan-2-ol
About This Item
Assay
97%
form
solid
mp
64-69 °C
functional group
hydroxyl
SMILES string
CC(C)(O)c1cccc(c1)B2OC(C)(C)C(C)(C)O2
InChI
1S/C15H23BO3/c1-13(2,17)11-8-7-9-12(10-11)16-18-14(3,4)15(5,6)19-16/h7-10,17H,1-6H3
InChI key
NNPQWBXXZHNLAW-UHFFFAOYSA-N
Application
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service