Skip to Content
Merck
All Photos(2)

Key Documents

720569

Sigma-Aldrich

4-(trans-2-Carboxyvinyl)phenylboronic acid

Synonym(s):

4-(2-Carboxy-(E)-ethenyl)benzeneboronic acid, 4-(E-2-Carboxyvinyl)phenylboronic acid

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$307.00

$307.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$307.00

About This Item

Empirical Formula (Hill Notation):
C9H9BO4
CAS Number:
Molecular Weight:
191.98
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

$307.00


Check Cart for Availability

Request a Bulk Order

form

solid

Quality Level

mp

187-193 °C

functional group

carboxylic acid

storage temp.

2-8°C

SMILES string

OB(O)c1ccc(\C=C\C(O)=O)cc1

InChI

1S/C9H9BO4/c11-9(12)6-3-7-1-4-8(5-2-7)10(13)14/h1-6,13-14H,(H,11,12)/b6-3+

InChI key

IEMLKNHGGSYOMP-ZZXKWVIFSA-N

Application

Reactant for:
  • Preparation of nickel thiocyanate pyridylphenylpropenate polymeric complex

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service