Skip to Content
Merck
All Photos(1)

Key Documents

719692

Sigma-Aldrich

cis-9-Octadecene-1-thiol

97%

Synonym(s):

Oleyl mercaptan

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)6CH2CH=CHCH2(CH2)6CH2SH
CAS Number:
Molecular Weight:
284.54
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
form:
liquid
Assay:
97%

Assay

97%

form

liquid

density

0.852 g/mL at 25 °C

storage temp.

−20°C

SMILES string

CCCCCCCC\C=C/CCCCCCCCS

InChI

1S/C18H36S/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19/h9-10,19H,2-8,11-18H2,1H3/b10-9-

InChI key

LJLJPBMSWIYGOA-KTKRTIGZSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

>230.0 °F - closed cup

Flash Point(C)

> 110 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service