719390
2-Pyridylboronic acid MIDA ester
Synonym(s):
2-Pyridinylboronic acid MIDA ester
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
form
solid
impurities
≤15 wt. % DMSO
storage temp.
2-8°C
SMILES string
CN1CC(=O)OB(OC(=O)C1)c2ccccn2
InChI
1S/C10H11BN2O4/c1-13-6-9(14)16-11(17-10(15)7-13)8-4-2-3-5-12-8/h2-5H,6-7H2,1H3
InChI key
OEKJACZNFHBYNV-UHFFFAOYSA-N
Application
MIDA boronates as stable boronic acid surrogates for classically challenging cross-couplings.
Suzuki Cross-Coupling with MIDA Boronates
Suzuki Cross-Coupling with MIDA Boronates
2-Pyridinylboronic acid MIDA ester represents the first and only example of an air-stable 2-pyridinyl boronate. This stable boronic acid surrogate is a powerful cross coupling partner, useful in the synthesis of pharmaceuticals, natural products, and complex molecules.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Organic letters, 12(10), 2314-2317 (2010-05-15)
A wide range of 2-pyridyl and other difficult-to-access heterocyclic N-methyliminodiacetic acid boronates can be readily prepared from the corresponding bromides via a new method involving direct transligation of 2-heterocyclic trialkoxyborate salts with N-methyliminodiacetic acid (MIDA) at elevated temperatures.
Journal of the American Chemical Society, 131(20), 6961-6963 (2009-05-02)
Many boronic acids, including 2-heterocyclic, vinyl, and cyclopropyl derivatives, are inherently unstable, which can limit their benchtop storage and/or efficient cross-coupling. We herein report the first general solution to this problem: in situ slow release of unstable boronic acids from
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service