Skip to Content
Merck
All Photos(1)

Key Documents

71916

Sigma-Aldrich

Sodium 2-propanethiolate

technical, ≥90.0% (RT)

Synonym(s):

2-Propanethiol sodium salt

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$214.00

$214.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 G
$214.00

About This Item

Linear Formula:
(CH3)2CHSNa
CAS Number:
Molecular Weight:
98.14
Beilstein:
3618618
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$214.00


Check Cart for Availability

Request a Bulk Order

grade

technical

Quality Level

Assay

≥90.0% (RT)

form

powder

SMILES string

CC(C)S[Na]

InChI

1S/C3H8S.Na/c1-3(2)4;/h3-4H,1-2H3;/q;+1/p-1

InChI key

NQHKCHWQDDWQJR-UHFFFAOYSA-M

Related Categories

Application

  • Biphenyl and bimesityl tetrasulfonic acid–new linker molecules for coordination polymers: This study discusses the use of Sodium 2-propanethiolate in the synthesis of new coordination polymers. The article details the process and the resulting chemical structures, providing insights into the potential applications of these materials in various fields (F Behler, MS Wickleder, J Christoffers - ARKIVOC, 2015).

Other Notes

Intermediate for the synthesis of various thiol esters and thio ethers. Synthesis of polymercapto-arenes from polyhalides via poly(isopropyl thio ethers)[1]

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L. Testaferri et al.
Synthesis, 751-751 (1983)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service