Skip to Content
Merck
All Photos(2)

Key Documents

716405

Sigma-Aldrich

2-Hydroxy-4-iodobenzoic acid

≥95%

Synonym(s):

4-Iodosalicylic acid

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$207.00

$207.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$207.00

About This Item

Empirical Formula (Hill Notation):
C7H5IO3
CAS Number:
Molecular Weight:
264.02
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$207.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥95%

form

powder

mp

229-234 °C

functional group

carboxylic acid
iodo

SMILES string

OC(=O)c1ccc(I)cc1O

InChI

1S/C7H5IO3/c8-4-1-2-5(7(10)11)6(9)3-4/h1-3,9H,(H,10,11)

InChI key

UQOZVZTUJNRMBV-UHFFFAOYSA-N

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H Goudonnet et al.
Comptes rendus des seances de la Societe de biologie et de ses filiales, 176(5), 707-715 (1982-01-01)
In the series of salicylic acids derivatives investigated 3,5-DIS has a large inductive effect of enzymes with respect to the metabolism of xenobiotics: rise in the rate of Cyt. P 450; increase of ethoxycoumarin deethylase, bilirubin glucuronosyl transferase and benzphetamine-N-demethylase

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service