Skip to Content
Merck
All Photos(1)

Key Documents

716324

Sigma-Aldrich

2-(Trifluoromethyl)piperazine

95%

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$152.00

$152.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
$152.00

About This Item

Empirical Formula (Hill Notation):
C5H9F3N2
CAS Number:
Molecular Weight:
154.13
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$152.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

95%

form

powder

mp

90-95 °C

functional group

fluoro

SMILES string

FC(F)(F)C1CNCCN1

InChI

1S/C5H9F3N2/c6-5(7,8)4-3-9-1-2-10-4/h4,9-10H,1-3H2

InChI key

YNEJOOCZWXGXDU-UHFFFAOYSA-N

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service